Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zhx3
Entrez Gene ID
311604
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
Tix1
Chromosome Location
Chr.3: 156759824 - 156868178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235474.2 XP_006235536.1 2 - 3 3663 195

View Full Product Details