Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zhx3
Entrez Gene ID
311604
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
Tix1
Chromosome Location
Chr.3: 156759824 - 156868178 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047097.1 NP_001040562.1 1 - 2 2846 85
XM_008762344.1 XP_008760566.1 3 - 4 3349 85
GenBank mRNA DQ017884.1 - 1 - 2 2846 85

View Full Product Details