The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004072.2 | NP_001004072.2 | 5 - 6 | 546 | 83 |
GenBank mRNA | BC079369.1 | - | 5 - 6 | 538 | 83 |
BC098897.1 | - | 5 - 6 | 546 | 83 | |
Z12158.1 | - | 5 - 6 | 565 | 83 |