Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnaja3
Entrez Gene ID
360481
Gene Name
DnaJ (Hsp40) homolog, subfamily A, member 3
Gene Aliases
Tid-1
Chromosome Location
Chr.10: 11059701 - 11085182 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038595.1 NP_001033684.1 5 - 6 783 75
NM_001038596.1 NP_001033685.1 5 - 6 783 75
XM_006245817.2 XP_006245879.1 5 - 6 823 75
GenBank mRNA AF516338.1 - 5 - 6 783 75
AY077460.1 - 5 - 6 783 75

View Full Product Details