Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glis2
Entrez Gene ID
302946
Gene Name
GLIS family zinc finger 2
Gene Aliases
-
Chromosome Location
Chr.10: 11154460 - 11174905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106978.1 NP_001100448.1 1 - 2 96 56
XM_006245801.2 XP_006245863.1 1 - 2 140 56
XM_008767487.1 XP_008765709.1 1 - 2 140 56
XM_008767488.1 XP_008765710.1 1 - 2 140 56

View Full Product Details