Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nagpa
Entrez Gene ID
360476
Gene Name
N-acetylglucosamine-1-phosphodiester alpha-N-acetylglucosaminidase
Gene Aliases
-
Chromosome Location
Chr.10: 10573191 - 10581516 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108265.1 NP_001101735.1 6 - 7 1166 64
GenBank mRNA BC168968.1 - 6 - 7 1152 64

View Full Product Details