The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105761.1 | NP_001099231.1 | 2 - 3 | 392 | 71 |
XM_008767480.1 | XP_008765702.1 | 2 - 3 | 401 | 71 |