Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srl
Entrez Gene ID
302948
Gene Name
sarcalumenin
Gene Aliases
-
Chromosome Location
Chr.10: 11240141 - 11284325 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008767510.1 XP_008765732.1 1 - 2 93 55

View Full Product Details