Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alg1
Entrez Gene ID
360475
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.10: 10539930 - 10550174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108264.1 NP_001101734.1 8 - 9 910 66

View Full Product Details