Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vhl
Entrez Gene ID
24874
Gene Name
von Hippel-Lindau tumor suppressor, E3 ubiquitin protein ligase
Gene Aliases
Vhlh
Chromosome Location
Chr.4: 145580869 - 145587835 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052801.1 NP_434688.1 2 - 3 476 92
GenBank mRNA S80345.1 - 2 - 3 417 92
U14746.1 - 2 - 3 476 92

View Full Product Details