The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108640.2 | NP_001102110.2 | 7 - 8 | 792 | 70 |
GenBank mRNA | BC168907.1 | - | 6 - 7 | 650 | 70 |