The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198972.1 | NP_945323.1 | 5 - 6 | 489 | 120 |
GenBank mRNA | AY456958.1 | - | 5 - 6 | 489 | 120 |