Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zxdc
Entrez Gene ID
362399
Gene Name
ZXD family zinc finger C
Gene Aliases
RGD1561296
Chromosome Location
Chr.4: 122282126 - 122314251 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729395.4 XP_002729441.3 3 - 4 1290 89

View Full Product Details