The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013926.2 | NP_001013948.2 | 3 - 4 | 391 | 129 |
XM_003749789.3 | XP_003749837.1 | 3 - 4 | 342 | 129 | |
XM_006236804.2 | XP_006236866.1 | 4 - 5 | 1048 | 129 | |
XM_006236805.2 | XP_006236867.1 | 4 - 5 | 543 | 129 | |
XM_006236808.2 | XP_006236870.1 | 4 - 5 | 413 | 129 | |
XM_008763062.1 | XP_008761284.1 | 5 - 6 | 1085 | 129 | |
GenBank mRNA | BC087060.1 | - | 4 - 5 | 406 | 129 |
CK475733.1 | - | 3 - 4 | 391 | 129 | |
CV112449.1 | - | 3 - 4 | 319 | 129 | |
FQ234910.1 | - | 3 - 4 | 348 | 129 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006236742.2 | XP_006236804.1 | - | 1048 | 129 |
XM_006236743.2 | XP_006236805.1 | - | 543 | 129 | |
XM_006236745.2 | XP_006236807.1 | - | 413 | 129 | |
XM_006236746.2 | XP_006236808.1 | - | 327 | 129 | |
XM_008763108.1 | XP_008761330.1 | - | 427 | 129 | |
XM_008763109.1 | XP_008761331.1 | - | 1085 | 129 |