Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hk2
Entrez Gene ID
25059
Gene Name
hexokinase 2
Gene Aliases
-
Chromosome Location
Chr.4: 113559396 - 113609897 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012735.2 NP_036867.1 13 - 14 2133 107
XM_006236711.2 XP_006236773.1 12 - 13 3202 107
GenBank mRNA AF027179.1 - 13 - 14 1937 107
M68971.1 - 13 - 14 2134 107
M68972.1 - 5 - 6 735 107

View Full Product Details