Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap25
Entrez Gene ID
500246
Gene Name
Rho GTPase activating protein 25
Gene Aliases
RGD1562105
Chromosome Location
Chr.4: 119249682 - 119327822 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109247.1 NP_001102717.1 8 - 9 1369 89
XM_006236838.2 XP_006236900.1 8 - 9 3528 89
XM_006236839.2 XP_006236901.1 7 - 8 1224 89

View Full Product Details