The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053645.1 | NP_446097.1 | 8 - 9 | 1068 | 88 |
XM_006236977.2 | XP_006237039.1 | 10 - 11 | 1459 | 88 | |
GenBank mRNA | AB056101.1 | - | 8 - 9 | 1068 | 88 |
AF324153.1 | - | 8 - 9 | 1068 | 88 |