Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp4r2
Entrez Gene ID
297486
Gene Name
protein phosphatase 4, regulatory subunit 2
Gene Aliases
-
Chromosome Location
Chr.4: 133285552 - 133323755 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106613.2 NP_001100083.2 4 - 5 604 69
XM_006236962.2 XP_006237024.1 4 - 5 678 69
GenBank mRNA BC167050.1 - 4 - 5 522 69
EV770270.1 - 4 - 5 624 69
EV776660.1 - 1 - 2 83 69

View Full Product Details