Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thnsl2
Entrez Gene ID
297332
Gene Name
threonine synthase-like 2 (S. cerevisiae)
Gene Aliases
RGD1309144, Tsh2
Chromosome Location
Chr.4: 98976559 - 98997192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009658.1 NP_001009658.1 4 - 5 625 99
XM_006236625.2 XP_006236687.1 - 755 99
XM_006236626.2 XP_006236688.1 - 755 99
XM_006236627.2 XP_006236689.1 - 561 99
GenBank mRNA BC088454.1 - 4 - 5 625 99

View Full Product Details