The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009658.1 | NP_001009658.1 | 7 - 8 | 1130 | 88 |
XM_006236625.2 | XP_006236687.1 | - | 1260 | 88 | |
XM_006236626.2 | XP_006236688.1 | - | 1260 | 88 | |
XM_006236627.2 | XP_006236689.1 | - | 1066 | 88 | |
GenBank mRNA | BC088454.1 | - | 7 - 8 | 1130 | 88 |