Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Capg
Entrez Gene ID
297339
Gene Name
capping protein (actin filament), gelsolin-like
Gene Aliases
-
Chromosome Location
Chr.4: 100402301 - 100419447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013086.1 NP_001013104.1 6 - 7 970 129
XM_006236661.2 XP_006236723.1 7 - 8 810 129
XM_006236662.2 XP_006236724.1 7 - 8 903 129
XM_006236663.2 XP_006236725.1 7 - 8 1037 129
XM_006236666.2 XP_006236728.1 6 - 7 757 129
GenBank mRNA BC079104.1 - 6 - 7 970 129

View Full Product Details