Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dqx1
Entrez Gene ID
680434
Gene Name
DEAQ box RNA-dependent ATPase 1
Gene Aliases
-
Chromosome Location
Chr.4: 113890198 - 113899350 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109407.1 NP_001102877.1 11 - 12 2063 77
XM_006236717.2 XP_006236779.1 11 - 12 2553 77
XM_008763030.1 XP_008761252.1 10 - 11 2738 77

Gene Symbol
LOC103692182
Entrez Gene ID
103692182
Gene Name
ATP-dependent RNA helicase DQX1-like
Gene Aliases
-
Chromosome Location
Chr.4: 114762816 - 114765195 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763119.1 XP_008761341.1 - 619 77

View Full Product Details