The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008562.1 | NP_001008562.1 | 5 - 6 | 1043 | 77 |
GenBank mRNA | BC079071.1 | - | 5 - 6 | 1043 | 77 |