Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinb5
Entrez Gene ID
116589
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 5
Gene Aliases
Maspin, PI-5, Pi5
Chromosome Location
Chr.13: 26903052 - 26923253 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057108.2 NP_476449.1 6 - 7 837 70
XM_008769405.1 XP_008767627.1 5 - 6 995 70

View Full Product Details