Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Serpinb2
Entrez Gene ID
60325
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 2
Gene Aliases
Pai2a
Chromosome Location
Chr.13: 27449895 - 27463015 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021696.2 NP_067728.1 4 - 5 568 158
XM_006249639.2 XP_006249701.1 5 - 6 754 158
XM_008769495.1 XP_008767717.1 5 - 6 651 158
GenBank mRNA X64563.1 - 4 - 5 569 158

View Full Product Details