Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Urb2
Entrez Gene ID
292087
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
RGD1307980
Chromosome Location
Chr.19: 56803708 - 56827017 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135708.1 NP_001129180.1 7 - 8 4235 85

View Full Product Details