The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108896.1 | NP_001102366.1 | 11 - 12 | 1401 | 80 |
GenBank mRNA | FQ228966.1 | - | 11 - 12 | 1319 | 80 |