Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Best2
Entrez Gene ID
364973
Gene Name
bestrophin 2
Gene Aliases
Vmd2l1
Chromosome Location
Chr.19: 26122869 - 26129619 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108895.1 NP_001102365.1 7 - 8 987 76
XM_006255283.2 XP_006255345.1 6 - 7 1121 76

View Full Product Details