The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001047847.1 | NP_001041312.1 | 8 - 9 | 867 | 60 |
GenBank mRNA | AY940050.1 | - | 8 - 9 | 867 | 60 |