Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc7
Entrez Gene ID
170906
Gene Name
zinc finger, DHHC-type containing 7
Gene Aliases
Serz-1, Serz1
Chromosome Location
Chr.19: 52733161 - 52750307 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133394.1 NP_596885.1 2 - 3 291 65
GenBank mRNA AY040615.1 - 2 - 3 291 65
AY886525.1 - 2 - 3 286 65
BC061769.1 - 1 - 2 67 65

View Full Product Details