The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108441.1 | NP_001101911.1 | 2 - 3 | 931 | 130 |
XM_006255282.2 | XP_006255344.1 | 2 - 3 | 898 | 130 | |
GenBank mRNA | BC159430.1 | - | 2 - 3 | 794 | 130 |
FQ227962.1 | - | 2 - 3 | 853 | 130 |