Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxw9
Entrez Gene ID
288921
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
-
Chromosome Location
Chr.19: 26173771 - 26180675 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081634.1 NP_001075103.1 8 - 9 1312 79
GenBank mRNA BC166590.1 - 5 - 6 1512 79

View Full Product Details