Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atg4b
Entrez Gene ID
316640
Gene Name
autophagy related 4B, cysteine peptidase
Gene Aliases
Apg4b
Chromosome Location
Chr.9: 100888450 - 100920123 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006245533.2 XP_006245595.1 3 - 4 309 158

View Full Product Details