Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fer
Entrez Gene ID
301737
Gene Name
fer (fps/fes related) tyrosine kinase
Gene Aliases
Fert2, Flk, Flk_retired
Chromosome Location
Chr.9: 111557802 - 111868583 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106928.1 NP_001100398.1 1 - 2 89 67

View Full Product Details