Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Agap1
Entrez Gene ID
316611
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
Centg2
Chromosome Location
Chr.9: 96718900 - 97034400 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108230.1 NP_001101700.1 4 - 5 373 67
XM_006245396.2 XP_006245458.2 6 - 7 4481 67
XM_006245397.2 XP_006245459.2 6 - 7 4481 67
XM_006245398.2 XP_006245460.2 6 - 7 4480 67
XM_006245400.2 XP_006245462.2 5 - 6 4468 67
XM_006245401.2 XP_006245463.2 6 - 7 4481 67
XM_006245402.2 XP_006245464.2 6 - 7 4481 67
XM_006245403.2 XP_006245465.2 5 - 6 4468 67

View Full Product Details