Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb3
Entrez Gene ID
680216
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 3
Gene Aliases
-
Chromosome Location
Chr.9: 95289918 - 95290931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109396.1 NP_001102866.1 1 - 1 963 152

View Full Product Details