Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thap4
Entrez Gene ID
363291
Gene Name
THAP domain containing 4
Gene Aliases
-
Chromosome Location
Chr.9: 100848597 - 100888107 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005564.1 NP_001005564.1 2 - 3 1252 99
GenBank mRNA BC081882.1 - 2 - 3 1252 99

View Full Product Details