Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxl17
Entrez Gene ID
316663
Gene Name
F-box and leucine-rich repeat protein 17
Gene Aliases
-
Chromosome Location
Chr.9: 110515558 - 110936631 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108235.1 NP_001101705.1 1 - 2 265 75

View Full Product Details