Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnpepl1
Entrez Gene ID
684035
Gene Name
arginyl aminopeptidase (aminopeptidase B)-like 1
Gene Aliases
-
Chromosome Location
Chr.9: 100080144 - 100089834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271177.1 NP_001258106.1 3 - 4 813 97

View Full Product Details