The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025012.1 | NP_001020183.1 | 7 - 8 | 1472 | 112 |
XM_006245604.1 | XP_006245666.1 | 6 - 7 | 1395 | 112 | |
XM_006245605.2 | XP_006245667.1 | 7 - 8 | 1891 | 112 | |
XM_008767377.1 | XP_008765599.1 | 7 - 8 | 5067 | 112 | |
XM_008767378.1 | XP_008765600.1 | 7 - 8 | 4727 | 112 | |
GenBank mRNA | BC082058.1 | - | 7 - 8 | 1472 | 112 |