Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdkn1a
Entrez Gene ID
114851
Gene Name
cyclin-dependent kinase inhibitor 1A
Gene Aliases
Cip1, UV96, Waf1
Chromosome Location
Chr.20: 6348422 - 6358864 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080782.3 NP_542960.1 3 - 3 1816 120
XM_006256128.2 XP_006256190.1 3 - 3 1881 120
GenBank mRNA BC100620.1 - 3 - 3 1816 120

View Full Product Details