Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbs
Entrez Gene ID
24250
Gene Name
cystathionine beta synthase
Gene Aliases
-
Chromosome Location
Chr.20: 10361987 - 10386663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012522.2 NP_036654.2 5 - 6 668 78
GenBank mRNA D01098.1 - 5 - 6 669 78
FQ211441.1 - 5 - 6 727 78
M88344.1 - 5 - 6 817 78
M88345.1 - 2 - 3 1081 78
M88346.1 - 4 - 5 563 78
M88347.1 - 4 - 5 563 78

View Full Product Details