Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab44
Entrez Gene ID
309649
Gene Name
RAB44, member RAS oncogene family
Gene Aliases
RGD1560150
Chromosome Location
Chr.20: 6363839 - 6410878 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256153.2 XP_006256215.1 9 - 10 2579 82

View Full Product Details