Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tapbp
Entrez Gene ID
25217
Gene Name
TAP binding protein (tapasin)
Gene Aliases
-
Chromosome Location
Chr.20: 5468056 - 5476007 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033098.2 NP_149089.1 1 - 2 62 94
GenBank mRNA AJ400732.1 - 1 - 2 51 94
BC062007.1 - 1 - 2 53 94
CB794019.1 - 1 - 2 121 94
FM054278.1 - 1 - 2 53 94
KC222917.1 - 1 - 2 39 94
KC222918.1 - 1 - 2 39 94
KC222919.1 - 1 - 2 39 94
KC222920.1 - 1 - 2 39 94
KC222921.1 - 1 - 2 39 94

View Full Product Details