The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017294.1 | NP_058990.1 | 3 - 4 | 364 | 146 |
GenBank mRNA | AF104402.1 | - | 3 - 4 | 364 | 146 |