Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryaa
Entrez Gene ID
24273
Gene Name
crystallin, alpha A
Gene Aliases
Acry-1, Crya1
Chromosome Location
Chr.20: 10438037 - 10442189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012534.4 NP_036666.2 1 - 2 246 121
GenBank mRNA BC126082.1 - 1 - 2 246 121
U47922.1 - 1 - 2 199 121

View Full Product Details