Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btbd9
Entrez Gene ID
294318
Gene Name
BTB (POZ) domain containing 9
Gene Aliases
-
Chromosome Location
Chr.20: 8899286 - 9253931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013073.1 NP_001013091.1 7 - 8 1362 141
XM_006256185.2 XP_006256247.1 7 - 8 1388 141
GenBank mRNA BC087068.1 - 7 - 8 1362 141

View Full Product Details