Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Brpf3
Entrez Gene ID
309647
Gene Name
bromodomain and PHD finger containing, 3
Gene Aliases
-
Chromosome Location
Chr.20: 6047792 - 6086099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107615.1 NP_001101085.1 6 - 7 2497 76
XM_006256129.2 XP_006256191.1 7 - 8 2937 76
XM_006256130.2 XP_006256192.1 7 - 8 2937 76
XM_006256131.2 XP_006256193.1 7 - 8 2646 76
XM_006256132.2 XP_006256194.1 7 - 8 2854 76

View Full Product Details