The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212529.1 | NP_997694.1 | 8 - 9 | 784 | 111 |
XM_006255966.2 | XP_006256028.1 | 7 - 8 | 862 | 111 | |
GenBank mRNA | BC091424.1 | - | 5 - 6 | 1096 | 111 |