Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab36
Entrez Gene ID
690407
Gene Name
RAB36, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.20: 14557511 - 14573948 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109589.2 NP_001103059.1 5 - 6 390 77
XM_006256342.2 XP_006256404.1 5 - 6 820 77
XM_008772925.1 XP_008771147.1 5 - 6 825 77
XM_008772926.1 XP_008771148.1 5 - 6 826 77
GenBank mRNA BC167029.1 - 5 - 6 390 77
FQ213589.1 - 4 - 5 316 77

View Full Product Details